Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133614.2 | NP_598298.1 | 6 | 631 | ||
XM_006240114.2 | XP_006240176.1 | 6 | 1957 | |||
GenBank mRNA | AJ289714.1 | - | 6 | 475 | ||
BC089099.1 | - | 6 | 631 | 7306738 |