Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hopx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133621.2 | NP_598305.2 | 2, 3 | 260 | ||
GenBank mRNA | AF474162.1 | - | 180 | |||
AF492685.1 | - | 2, 3 | 260 | |||
BC070950.1 | - | 1, 2 | 171 | 7098349 | ||
FQ212752.1 | - | 2, 3 | 257 | |||
FQ220854.1 | - | 2, 3 | 259 | |||
FQ221225.1 | - | 2, 3 | 275 | |||
FQ222004.1 | - | 2, 3 | 250 | |||
FQ222724.1 | - | 2, 3 | 192 | |||
FQ229425.1 | - | 2, 3 | 245 | |||
FQ229457.1 | - | 2, 3 | 237 | |||
FQ229504.1 | - | 2, 3 | 237 | |||
FQ229663.1 | - | 2, 3 | 249 | |||
FQ229876.1 | - | 2, 3 | 237 | |||
FQ229892.1 | - | 2, 3 | 275 | |||
FQ230105.1 | - | 1, 2 | 253 | |||
FQ230629.1 | - | 2, 3 | 258 |