Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hopx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133621.2 | NP_598305.2 | 3 | 479 | ||
GenBank mRNA | AF474162.1 | - | 398 | |||
AF492685.1 | - | 3 | 479 | |||
BC070950.1 | - | 2 | 390 | 7098349 | ||
FQ212752.1 | - | 3 | 476 | |||
FQ220854.1 | - | 3 | 478 | |||
FQ221225.1 | - | 3 | 494 | |||
FQ222004.1 | - | 3 | 469 | |||
FQ222351.1 | - | 3 | 444 | |||
FQ222724.1 | - | 3 | 411 | |||
FQ229425.1 | - | 3 | 464 | |||
FQ229457.1 | - | 3 | 456 | |||
FQ229504.1 | - | 3 | 456 | |||
FQ229663.1 | - | 3 | 468 | |||
FQ229876.1 | - | 3 | 456 | |||
FQ229892.1 | - | 3 | 494 | |||
FQ230105.1 | - | 2 | 472 | |||
FQ230629.1 | - | 3 | 477 |