Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003218.3 | NP_003209.2 | 6 | 864 | ||
NM_017489.2 | NP_059523.2 | 6 | 864 | |||
XM_005251291.3 | XP_005251348.1 | 6 | 862 | |||
XM_005251292.3 | XP_005251349.1 | 6 | 862 | |||
XM_011517582.2 | XP_011515884.1 | 7 | 929 | |||
XM_017013792.1 | XP_016869281.1 | 7 | 762 | |||
GenBank mRNA | AF003001.1 | - | 6 | 854 | ||
AK292529.1 | - | 6 | 862 | |||
BC029378.1 | - | 6 | 847 | 4724068 | ||
BC071975.1 | - | 6 | 840 | |||
U40705.1 | - | 6 | 854 | |||
U74382.1 | - | 6 | 839 |