Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001156.4 | NP_001147.1 | 8 | 729 | ||
NM_001320879.1 | NP_001307808.1 | 7 | 674 | |||
NM_001320880.1 | NP_001307809.1 | 8 | 740 | |||
NM_004034.3 | NP_004025.1 | 9 | 795 | |||
XM_017016162.1 | XP_016871651.1 | 9 | 831 | |||
XM_017016163.1 | XP_016871652.1 | 8 | 765 | |||
GenBank mRNA | AB062429.1 | - | 8 | 687 | ||
AK222552.1 | - | 8 | 697 | |||
AK295319.1 | - | 8 | 606 | |||
AK300086.1 | - | 6 | 496 | |||
AK301681.1 | - | 8 | 483 | |||
AK307772.1 | - | 7 | 548 | |||
AK309141.1 | - | 8 | 491 | |||
AK312447.1 | - | 9 | 768 | |||
AK313024.1 | - | 8 | 672 | |||
BC002632.2 | - | 8 | 689 | 3610541 | ||
BT007187.1 | - | 7 | 654 | |||
BX361177.2 | - | 7 | 640 | |||
CR407686.1 | - | 7 | 654 | |||
J04543.1 | - | 8 | 714 |