Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318230.1 | NP_001305159.1 | 2 | 573 | ||
NM_006285.2 | NP_006276.2 | 2 | 573 | |||
GenBank mRNA | AK298907.1 | - | 2 | 111 | ||
BC037561.1 | - | 2 | 168 | |||
BC038448.1 | - | 2 | 547 | 5170888 | ||
BC067130.1 | - | 2 | 584 | 6153652 | ||
D50863.1 | - | 2 | 509 |