Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001063.3 | NP_001054.1 | 16 | 2344 | ||
XM_017007089.1 | XP_016862578.1 | 22 | 3694 | |||
XM_017007090.1 | XP_016862579.1 | 22 | 4687 | |||
GenBank mRNA | AK126941.1 | - | 2 | 3036 | ||
AK222755.1 | - | 16 | 2084 | |||
AK295334.1 | - | 16 | 2001 | |||
AK295419.1 | - | 15 | 1911 | |||
AK303753.1 | - | 16 | 2024 | |||
BC059367.1 | - | 16 | 2082 | 5264299 | ||
BX537660.1 | - | 2 | 1378 | |||
BX648533.1 | - | 16 | 1861 | |||
CR936810.1 | - | 21 | 3766 | |||
DQ923758.1 | - | 16 | 2075 | |||
GQ472199.1 | - | 16 | 2082 | |||
M12525.1 | - | 7 | 774 | |||
M12530.1 | - | 16 | 2066 | |||
M26641.1 | - | 1 | 131 | |||
S95936.1 | - | 16 | 2115 |