Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134358.1 | NP_599185.1 | 7 | 1048 | ||
GenBank mRNA | BC083694.1 | - | 7 | 1445 | ||
BC158686.1 | - | 7 | 1048 | 8374870 | ||
FQ214448.1 | - | 7 | 1024 | |||
FQ220404.1 | - | 7 | 1001 | |||
FQ220639.1 | - | 7 | 1064 | |||
FQ220740.1 | - | 7 | 1049 | |||
FQ220961.1 | - | 7 | 1049 | |||
FQ229287.1 | - | 7 | 1051 | |||
FQ229512.1 | - | 7 | 1064 | |||
FQ230125.1 | - | 7 | 1001 |