Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001063.3 | NP_001054.1 | 10, 11 | 1615 | ||
XM_017007089.1 | XP_016862578.1 | 16, 17 | 2965 | |||
XM_017007090.1 | XP_016862579.1 | 16, 17 | 3958 | |||
GenBank mRNA | AK222755.1 | - | 10, 11 | 1355 | ||
AK295334.1 | - | 10, 11 | 1272 | |||
AK295419.1 | - | 9, 10 | 1182 | |||
AK303753.1 | - | 10, 11 | 1295 | |||
BC059367.1 | - | 10, 11 | 1353 | 5264299 | ||
BX648533.1 | - | 10, 11 | 1132 | |||
CR936810.1 | - | 15, 16 | 3037 | |||
DQ923758.1 | - | 10, 11 | 1346 | |||
GQ472199.1 | - | 10, 11 | 1353 | |||
M12525.1 | - | 1, 2 | 45 | |||
M12530.1 | - | 10, 11 | 1337 | |||
S95936.1 | - | 10, 11 | 1386 |