Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134360.1 | NP_599187.1 | 7, 8 | 663 | ||
XM_006235511.2 | XP_006235573.1 | 7, 8 | 725 | |||
XM_008762465.1 | XP_008760687.1 | 6, 7 | 663 | |||
GenBank mRNA | BC097949.1 | - | 7, 8 | 663 | 7387440 | |
FQ226992.1 | - | 7, 8 | 646 | |||
FQ231420.1 | - | 7, 8 | 666 |