Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134367.1 | NP_599194.1 | 8 | 1021 | ||
XM_008770813.1 | XP_008769035.1 | 8 | 1204 | |||
XM_008770814.1 | XP_008769036.1 | 8 | 1203 | |||
XM_008770815.1 | XP_008769037.1 | 8 | 1638 | |||
XM_008770816.1 | XP_008769038.1 | 9 | 1015 | |||
XM_008770817.1 | XP_008769039.1 | 8 | 1552 | |||
GenBank mRNA | BC079027.1 | - | 8 | 1021 | 7114037 | |
GQ376198.1 | - | 8 | 930 | |||
GQ376199.1 | - | 8 | 930 |