Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134368.1 | NP_599195.1 | 15 | 2577 | ||
XM_006242596.2 | XP_006242658.1 | 15 | 2574 | |||
XM_006242597.2 | XP_006242659.1 | 15 | 2574 | |||
XM_006242598.2 | XP_006242660.1 | 15 | 2574 | |||
XM_006242599.2 | XP_006242661.1 | 15 | 2574 | |||
XM_006242600.2 | XP_006242662.1 | 15 | 2574 | |||
XM_006242601.2 | XP_006242663.1 | 15 | 2574 | |||
GenBank mRNA | AJ504723.1 | - | 14 | 2255 |