Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134378.2 | NP_599205.1 | 11, 12 | 1667 | ||
XM_008763486.1 | XP_008761708.1 | 11, 12 | 2239 | |||
XM_008763487.1 | XP_008761709.1 | 11, 12 | 2393 | |||
XM_008763488.1 | XP_008761710.1 | 11, 12 | 1848 | |||
GenBank mRNA | AF230072.1 | - | 11, 12 | 1668 |