Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102549542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761485.1 | XP_008759707.1 | 4, 5 | 606 |
Gene Symbol: Elovl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134383.2 | NP_599210.1 | 4, 5 | 463 | ||
GenBank mRNA | AB071986.1 | - | 4, 5 | 463 |