Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134391.1 | NP_599218.1 | 2 | 336 | ||
GenBank mRNA | AF329091.1 | - | 2 | 96 | ||
AJ309150.1 | - | 2 | 336 | |||
AY621353.1 | - | 2 | 336 |