Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135169.2 | NP_001128641.1 | 5 | 378 | ||
NM_134395.2 | NP_599222.2 | 4 | 591 | |||
XM_006253777.2 | XP_006253839.1 | 5 | 581 | |||
GenBank mRNA | AF083396.1 | - | 4 | 599 | ||
BC158854.1 | - | 4 | 272 | 7936967 | ||
CB744788.1 | - | 5 | 432 | |||
FQ211234.1 | - | 5 | 368 | |||
FQ217516.1 | - | 5 | 368 | |||
FQ218209.1 | - | 5 | 374 | |||
FQ223586.1 | - | 5 | 358 | |||
FQ223742.1 | - | 5 | 368 | |||
FQ224495.1 | - | 5 | 374 | |||
FQ225707.1 | - | 5 | 348 | |||
FQ230083.1 | - | 5 | 396 | |||
FQ230323.1 | - | 5 | 377 | |||
FQ231364.1 | - | 5 | 358 | |||
FQ233821.1 | - | 5 | 366 |