Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnttip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134400.2 | NP_599227.2 | 8 | 623 | ||
XM_006235512.2 | XP_006235574.1 | 8 | 642 | |||
XM_006235513.2 | XP_006235575.1 | 8 | 591 | |||
GenBank mRNA | AF348701.1 | - | 8 | 592 | ||
BC100640.1 | - | 8 | 623 | 7929876 |