Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001157.2 | NP_001148.1 | 8 | 1159 | ||
NM_001278407.1 | NP_001265336.1 | 10 | 1138 | |||
NM_001278408.1 | NP_001265337.1 | 10 | 1746 | |||
NM_001278409.1 | NP_001265338.1 | 10 | 1477 | |||
NM_145868.1 | NP_665875.1 | 9 | 1208 | |||
NM_145869.1 | NP_665876.1 | 10 | 1404 | |||
XM_005269741.4 | XP_005269798.1 | 6 | 1236 | |||
XM_005269742.1 | XP_005269799.1 | 8 | 1162 | |||
XM_006717813.1 | XP_006717876.1 | 9 | 1173 | |||
XM_006717814.3 | XP_006717877.1 | 9 | 1162 | |||
XM_011539735.1 | XP_011538037.1 | 10 | 1687 | |||
XM_011539736.2 | XP_011538038.1 | 10 | 1687 | |||
GenBank mRNA | AB209770.1 | - | 9 | 1153 | ||
AJ278463.1 | - | 8 | 1158 | |||
AJ278464.1 | - | 9 | 1207 | |||
AJ278465.1 | - | 10 | 1403 | |||
AK000567.1 | - | 8 | 1158 | |||
AK056107.1 | - | 10 | 1138 | |||
AK126287.1 | - | 10 | 1746 | |||
AK222918.1 | - | 8 | 1121 | |||
AK298362.1 | - | 9 | 969 | |||
AK300509.1 | - | 6 | 1271 | |||
AK301047.1 | - | 10 | 1462 | |||
BC007564.2 | - | 9 | 994 | 2989058 | ||
BT019934.1 | - | 7 | 921 | |||
CR450323.1 | - | 7 | 921 | |||
L19605.1 | - | 8 | 1099 |