Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003223.2 | NP_003214.1 | 5 | 807 | ||
XM_011522633.2 | XP_011520935.1 | 5 | 943 | |||
XM_011522635.2 | XP_011520937.1 | 5 | 623 | |||
GenBank mRNA | AB209146.1 | - | 4 | 4949 | ||
AK309301.1 | - | 4 | 1248 | |||
BC010576.1 | - | 5 | 696 | 4181538 | ||
BU159415.1 | - | 5 | 761 | |||
CR541726.1 | - | 5 | 553 | |||
S73885.1 | - | 5 | 811 | |||
X57435.1 | - | 5 | 502 |