Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134468.1 | NP_604463.1 | 7 | 733 | ||
XM_006236999.2 | XP_006237061.1 | 8 | 1402 | |||
XM_006237000.2 | XP_006237062.1 | 8 | 1398 | |||
XM_006237001.2 | XP_006237063.1 | 7 | 1265 | |||
XM_006237002.2 | XP_006237064.1 | 7 | 1192 | |||
XM_006237003.2 | XP_006237065.1 | 7 | 1191 | |||
GenBank mRNA | BC071177.1 | - | 7 | 668 | 7098592 | |
FQ213485.1 | - | 696 | ||||
L24907.1 | - | 7 | 707 | |||
L26288.1 | - | 7 | 733 |