Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085369.1 | NP_001078838.1 | 5 | 421 | ||
NM_134472.3 | NP_608302.2 | 4 | 306 | |||
XM_006238632.2 | XP_006238694.1 | 5 | 470 | |||
XM_006238633.1 | XP_006238695.1 | 4 | 234 | |||
XM_006238634.2 | XP_006238696.1 | 4 | 766 | |||
XM_006238635.2 | XP_006238697.1 | 5 | 643 | |||
XM_006238636.2 | XP_006238698.1 | 5 | 639 | |||
GenBank mRNA | CO575506.1 | - | 4 | 229 |