Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138522.1 | NP_612531.1 | 3 | 223 | ||
XM_006250721.2 | XP_006250783.1 | 3 | 453 | |||
GenBank mRNA | D21095.1 | - | 3 | 223 | ||
D87927.1 | - | 3 | 223 |