Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccrn4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138526.1 | NP_612535.1 | 3 | 1066 | ||
GenBank mRNA | AF199495.1 | - | 2 | 623 |