Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138542.2 | NP_612551.2 | 4 | 711 | ||
XM_008762077.1 | XP_008760299.1 | 4 | 1018 | |||
GenBank mRNA | AF097887.1 | - | 3 | 666 | ||
BC086990.1 | - | 4 | 711 | 7135643 |