Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231862.2 | XP_006231924.1 | 11 | 3272 | ||
XM_006231863.2 | XP_006231925.1 | 11 | 3076 | |||
XM_008760695.1 | XP_008758917.1 | 15 | 3623 | |||
XM_008760696.1 | XP_008758918.1 | 14 | 3518 | |||
XM_008760697.1 | XP_008758919.1 | 14 | 3504 | |||
XM_008760698.1 | XP_008758920.1 | 10 | 2938 |