Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138549.1 | NP_612558.1 | 3 | 242 | ||
XM_006255216.2 | XP_006255278.1 | 2 | 377 | |||
XM_006255217.2 | XP_006255279.1 | 2 | 289 | |||
XM_006255218.2 | XP_006255280.1 | 3 | 247 | |||
GenBank mRNA | BC059115.1 | - | 3 | 205 | 6920522 | |
EU000473.1 | - | 3 | 205 | |||
FQ212009.1 | - | 3 | 228 | |||
FQ215654.1 | - | 2 | 299 | |||
FQ215778.1 | - | 1 | 109 | |||
FQ216428.1 | - | 2 | 294 | |||
FQ219100.1 | - | 3 | 182 | |||
FQ220726.1 | - | 3 | 191 | |||
S45663.1 | - | 3 | 242 |
Gene Symbol: RGD1560015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574311.6 | XP_574311.1 | 1 | 143 |