Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003954.2 | NP_001003954.1 | 7, 8 | 691 | ||
NM_004306.3 | NP_004297.2 | 6, 7 | 568 | |||
GenBank mRNA | AJ306450.1 | - | 7, 8 | 667 | ||
AK223374.1 | - | 6, 7 | 567 | |||
AK290281.1 | - | 7, 8 | 677 | |||
BC125158.1 | - | 6, 7 | 538 | 40120955 | ||
CR541838.1 | - | 5, 6 | 476 | |||
Z11502.1 | - | 6, 7 | 555 |