Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138895.1 | NP_620250.1 | 133 | |||
GenBank mRNA | BC060312.1 | - | 172 | 6919609 | ||
BC070919.1 | - | 142 | 7104905 | |||
BC078726.1 | - | 1 | 72 | |||
D16554.1 | - | 133 |
Gene Symbol: LOC100360645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727721.3 | XP_002727767.1 | 1 | 72 |
Gene Symbol: LOC100910768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750018.3 | XP_003750066.1 | 1 | 138 |
Gene Symbol: Uba52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U75407.1 | - | 107 |