Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206855.1 | NP_001193784.1 | 4 | 510 | ||
NM_003227.3 | NP_003218.2 | 7 | 930 | |||
XM_005250553.4 | XP_005250610.1 | 8 | 1236 | |||
XM_017012573.1 | XP_016868062.1 | 8 | 1038 | |||
GenBank mRNA | AF067864.1 | - | 7 | 916 | ||
AF085928.1 | - | 4 | 373 | |||
AK000421.1 | - | 2 | 144 | |||
BC142630.1 | - | 7 | 916 | 40147690 | ||
BX384750.2 | - | 4 | 510 |