Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100477.1 | NP_001093947.1 | 5 | 774 | ||
GenBank mRNA | D50580.1 | - | 5 | 766 | ||
FQ218265.1 | - | 5 | 814 |