Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000660.5 | NP_000651.3 | 4 | 1589 | ||
XM_011527242.1 | XP_011525544.1 | 4 | 1582 | |||
GenBank mRNA | AK291907.1 | - | 4 | 1078 | ||
AK307742.1 | - | 4 | 1274 | |||
AY820829.1 | - | 3 | 203 | |||
BC000125.1 | - | 4 | 1146 | 3351664 | ||
BC001180.1 | - | 4 | 1146 | 3356605 | ||
BC022242.1 | - | 4 | 1069 | 4399762 | ||
BT007245.1 | - | 4 | 700 | |||
X02812.1 | - | 4 | 1544 |