Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003954.2 | NP_001003954.1 | 12 | 1073 | ||
NM_004306.3 | NP_004297.2 | 11 | 950 | |||
GenBank mRNA | AJ306450.1 | - | 12 | 1049 | ||
AK223374.1 | - | 11 | 949 | |||
AK290281.1 | - | 12 | 1059 | |||
BC125158.1 | - | 11 | 920 | 40120955 | ||
CR541838.1 | - | 10 | 858 | |||
Z11502.1 | - | 11 | 937 |