Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139101.2 | NP_620801.1 | 2 | 383 | ||
GenBank mRNA | U78090.1 | - | 383 |