Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000358.2 | NP_000349.1 | 7 | 960 | ||
GenBank mRNA | AB209598.1 | - | 1 | 3722 | ||
AK093916.1 | - | 7 | 815 | |||
AK094055.1 | - | 8 | 891 | |||
AK094581.1 | - | 7 | 904 | |||
AK222833.1 | - | 7 | 868 | |||
AK298603.1 | - | 2 | 180 | |||
AK303643.1 | - | 7 | 1192 | |||
BC000097.2 | - | 7 | 853 | 3506051 | ||
BC004972.1 | - | 7 | 848 | 2958878 | ||
BT009820.1 | - | 7 | 799 | |||
KU178357.1 | - | 7 | 799 | |||
M77349.1 | - | 7 | 846 |