Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139114.3 | NP_620814.1 | 4 | 528 | ||
GenBank mRNA | BC078724.1 | - | 3 | 467 | 7125608 | |
FQ210520.1 | - | 4 | 483 | |||
FQ217417.1 | - | 4 | 484 | |||
FQ217514.1 | - | 4 | 487 | |||
FQ218767.1 | - | 4 | 483 | |||
FQ221301.1 | - | 4 | 496 | |||
FQ221436.1 | - | 4 | 485 | |||
FQ221541.1 | - | 4 | 484 | |||
FQ221627.1 | - | 4 | 487 | |||
FQ221775.1 | - | 4 | 484 | |||
FQ221816.1 | - | 4 | 484 | |||
FQ222173.1 | - | 4 | 484 | |||
FQ222227.1 | - | 4 | 487 | |||
FQ222322.1 | - | 4 | 483 | |||
FQ222450.1 | - | 4 | 497 | |||
FQ222725.1 | - | 4 | 484 | |||
FQ224390.1 | - | 4 | 483 | |||
FQ227511.1 | - | 4 | 498 | |||
FQ228660.1 | - | 3 | 472 | |||
FQ230911.1 | - | 4 | 487 | |||
X78167.1 | - | 3 | 465 |