Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139185.1 | NP_631924.1 | 4 | 475 | ||
XM_006228384.2 | XP_006228446.1 | 3 | 829 | |||
XM_006228385.2 | XP_006228447.1 | 3 | 273 | |||
GenBank mRNA | L35921.1 | - | 4 | 475 |