Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139258.1 | NP_640351.1 | 3 | 343 | ||
XM_006234723.2 | XP_006234785.1 | 3 | 381 | |||
XM_006234724.2 | XP_006234786.1 | 3 | 384 | |||
XM_008762079.1 | XP_008760301.1 | 3 | 382 | |||
GenBank mRNA | AF506761.1 | - | 3 | 343 |