Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139259.2 | NP_640352.1 | 4 | 430 | ||
XM_006243921.2 | XP_006243983.1 | 4 | 434 | |||
GenBank mRNA | AF497263.1 | - | 4 | 434 | ||
AF497264.1 | - | 4 | 282 | |||
AF534395.1 | - | 3 | 246 |