Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105756.1 | NP_001099226.1 | 1 | 248 | ||
GenBank mRNA | AF226629.1 | - | 1 | 194 |