Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037093.2 | NP_001032170.2 | 39 | 9605 | ||
XM_006236023.2 | XP_006236085.1 | 41 | 9726 | |||
XM_006236025.2 | XP_006236087.1 | 40 | 9735 | |||
XM_008762685.1 | XP_008760907.1 | 42 | 9840 | |||
XM_008762686.1 | XP_008760908.1 | 41 | 9801 | |||
GenBank mRNA | DQ228948.2 | - | 10 | 1818 |