Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000359.2 | NP_000350.1 | 10 | 1604 | ||
GenBank mRNA | AK291350.1 | - | 10 | 1571 | ||
AK299436.1 | - | 10 | 1373 | |||
AK301652.1 | - | 4 | 502 | |||
AK315819.1 | - | 10 | 1604 | |||
AK315828.1 | - | 10 | 1564 | |||
AK315843.1 | - | 10 | 1571 | |||
BC034699.1 | - | 10 | 1555 | 4747045 | ||
D90287.1 | - | 10 | 1560 | |||
M55183.1 | - | 10 | 1533 | |||
M62925.1 | - | 10 | 1566 | |||
X57974.1 | - | 4 | 358 |