Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323316.1 | NP_001310245.1 | 8 | 1148 | ||
NM_001323317.1 | NP_001310246.1 | 6 | 922 | |||
NM_001323318.1 | NP_001310247.1 | 6 | 985 | |||
NM_004613.3 | NP_004604.2 | 7 | 1165 | |||
NM_198951.2 | NP_945189.1 | 7 | 1165 | |||
XM_011529028.1 | XP_011527330.1 | 8 | 1110 | |||
GenBank mRNA | AK058031.1 | - | 7 | 933 | ||
AK291714.1 | - | 7 | 1006 | |||
AK295775.1 | - | 6 | 763 | |||
AK300292.1 | - | 6 | 826 | |||
AK314618.1 | - | 7 | 1006 | |||
AY675221.1 | - | 6 | 934 | |||
BC003551.1 | - | 7 | 1018 | 3544757 | ||
EU794665.1 | - | 7 | 1034 | |||
M55153.1 | - | 7 | 1069 | |||
M98478.1 | - | 7 | 1066 |