Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323316.1 | NP_001310245.1 | 10 | 1446 | ||
NM_001323317.1 | NP_001310246.1 | 8 | 1220 | |||
NM_001323318.1 | NP_001310247.1 | 8 | 1283 | |||
NM_004613.3 | NP_004604.2 | 9 | 1463 | |||
NM_198951.2 | NP_945189.1 | 9 | 1463 | |||
XM_011529028.1 | XP_011527330.1 | 10 | 1408 | |||
GenBank mRNA | AK058031.1 | - | 9 | 1231 | ||
AK291714.1 | - | 9 | 1304 | |||
AK295775.1 | - | 8 | 1061 | |||
AK300292.1 | - | 8 | 1124 | |||
AK314618.1 | - | 9 | 1304 | |||
AL512703.1 | - | 1 | 1006 | |||
AY675221.1 | - | 8 | 1232 | |||
BC003551.1 | - | 9 | 1316 | 3544757 | ||
BQ938957.1 | - | 1 | 24 | |||
EU794665.1 | - | 9 | 1332 | |||
M55153.1 | - | 9 | 1367 | |||
M98478.1 | - | 9 | 1364 |