Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144743.1 | NP_653344.1 | 314 | |||
GenBank mRNA | AY034877.1 | - | 314 |