Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003245.3 | NP_003236.3 | 10 | 1685 | ||
GenBank mRNA | AK290324.1 | - | 10 | 1649 | ||
AK291351.1 | - | 10 | 1649 | |||
AK315236.1 | - | 10 | 1649 | |||
BC109075.1 | - | 10 | 1611 | 40034239 | ||
BC109076.1 | - | 10 | 1611 | 40034240 | ||
BG675531.1 | - | 1 | 185 | |||
BX284087.1 | - | 1 | 185 | |||
L10386.1 | - | 10 | 1629 |