Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145082.2 | NP_659550.2 | 11 | 1689 | ||
GenBank mRNA | U53513.1 | - | 2 | 266 |