Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retsat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145084.1 | NP_659552.1 | 9 | 1491 | ||
GenBank mRNA | AF465614.1 | - | 9 | 1491 |