Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113570.1 | NP_001107042.1 | 5, 6 | 724 | ||
GenBank mRNA | AF391798.1 | - | 5, 6 | 485 |