Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003246.3 | NP_003237.2 | 19 | 3235 | ||
XM_011521971.1 | XP_011520273.1 | 18 | 3057 | |||
GenBank mRNA | AB209912.1 | - | 19 | 3221 | ||
AK291639.1 | - | 19 | 3235 | |||
AK304754.1 | - | 19 | 2981 | |||
AK310820.1 | - | 9 | 1770 | |||
BC136469.1 | - | 19 | 3209 | 9020458 | ||
BC136470.1 | - | 19 | 3209 | 9020459 | ||
X04665.1 | - | 19 | 3131 | |||
X14787.1 | - | 19 | 3167 |