Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003247.3 | NP_003238.2 | 19, 20 | 3318 | ||
GenBank mRNA | AK292429.1 | - | 19, 20 | 3275 | ||
BC146676.1 | - | 18, 19 | 3125 | 40148868 | ||
BC150175.1 | - | 18, 19 | 3129 | 40147958 | ||
BX641023.1 | - | 5, 6 | 773 | |||
L12350.1 | - | 19, 20 | 3237 | |||
M81339.1 | - | 8, 9 | 1322 |